Learn More
Selleck Chemical LLC Methylene Blue S4535-500mg
SDP
Catalog No. 503111023
Encompass_Preferred
Methylene Blue (Basic Blue 9 Tetramethylthionine chloride methylthioninium chloride CI-52015) is used as a dye in chromoendoscopy It Inhibits tau filament formation with IC50 of 1 9 M Also it inhibits soluble guanylyl cyclase
Catalog No. 50-311-1023 Supplier Selleck Chemical LLC Supplier No. S4535500MG
Methylene Blue (Basic Blue 9 Tetramethylthionine chloride methylthioninium chloride CI-52015) is used as a dye in chromoendoscopy It Inhibits tau filament formation with IC50 of 1 9 M Also it inhibits soluble guanylyl cyclase