Learn More
Sigma Aldrich Fine Chemicals Biosciences Methylene blue, 122965-43-9, MFCD00150006, 25 G
Catalog No. NC0772734
Encompass
Empirical Formula (Hill Notation): C16H18ClN3S · xH2O, Molecular Weight: 319.85 (anhydrous basis), Synonym: 3,7-bis(Dimethylamino)phenazathionium chloride, Basic Blue 9, Tetramethylthionine chloride. Methylene blue has been used: to stain vaginal cells, for the preparation of protein aqueous ink, and to evaluate cell survival by staining.
Catalog No. NC0772734 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M914025G
Empirical Formula (Hill Notation): C16H18ClN3S · xH2O, Molecular Weight: 319.85 (anhydrous basis), Synonym: 3,7-bis(Dimethylamino)phenazathionium chloride, Basic Blue 9, Tetramethylthionine chloride. Methylene blue has been used: to stain vaginal cells, for the preparation of protein aqueous ink, and to evaluate cell survival by staining.