Learn More
Volu Sol New Methylene Blue, 1 oz / 30 mL, Volu-Sol™
SDP
Supplier:  Volu Sol VMB030
Encompass_Preferred
Used as a blood and reticulocyte staining solution.  When mixed with whole blood, the RNA and DNA stain deep blue
Catalog No. NC9538424