Learn More
Molecular Research Center Methylene Blue Stain, 500 mL
SDP
Catalog No. NC9873714
Encompass
Methylene Blue Stain is an aqueous solution of methylene blue designed for quantitative and qualitative assessment of RNA and DNA immobilized on hybridization membranes. It can be used to verify the amount of RNA on hybridization membranes in northern blotting.
Catalog No. NC9873714 Supplier Molecular Research Center Supplier No. MB 119
Methylene Blue Stain is an aqueous solution of methylene blue designed for quantitative and qualitative assessment of RNA and DNA immobilized on hybridization membranes. It can be used to verify the amount of RNA on hybridization membranes in northern blotting.