Learn More
Apexbio Technology LLC Methylene Blue 61-73-4 500mg
SDP
Catalog No. 5000090704
Encompass_Preferred
Methylene blue (CAS 61-73-4) also known as methylthioninium chloride is a heterocyclic aromatic compound that functions as an oxidation-reduction agent It acts by facilitating electron transfer particularly in the reduction of methemoglobin to hemoglobin In biomedical research methylene blue has been extensively studied for its ability to inhibit Tau protein aggregation a process implicated in neurodegenerative disorders such as Alzheimer s disease Additionally it has been investigated for antiparasitic properties in malaria and explored as a urinary tract antiseptic Its multi-modal actions underpin its utility in diverse preclinical and clinical research contexts
Catalog No. 50-000-90704 Supplier Apexbio Technology LLC Supplier No. C6646500
Methylene blue (CAS 61-73-4) also known as methylthioninium chloride is a heterocyclic aromatic compound that functions as an oxidation-reduction agent It acts by facilitating electron transfer particularly in the reduction of methemoglobin to hemoglobin In biomedical research methylene blue has been extensively studied for its ability to inhibit Tau protein aggregation a process implicated in neurodegenerative disorders such as Alzheimer s disease Additionally it has been investigated for antiparasitic properties in malaria and explored as a urinary tract antiseptic Its multi-modal actions underpin its utility in diverse preclinical and clinical research contexts