Learn More
Medchemexpress LLC Methylene blue | 61-73-4 | 95.0% | C16H18ClN3S | 1 G
SDP
Catalog No. 5000355805
Encompass_Preferred
Methylene blue is an inhibitor of guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). It functions as a vasopressor and is frequently employed as a dye in various medical procedures. This compound is a REDOX cycling compound capable of crossing the blood-brain barrier and has shown to reduce cerebral edema, attenuate microglial activation, and diminish neuroinflammation.
  • Inhibits guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS)
  • Functions as a vasopressor
  • Employed as a dye in medical procedures
  • Influences the nitric oxide synthase/guanylate cyclase signaling pathway
  • REDOX cycling compound
  • Capable of crossing the blood-brain barrier
  • Acts as a Tau aggregation inhibitor
  • Reduces cerebral edema
  • Attenuates microglial activation
  • Diminishes neuroinflammation
Catalog No. 50-003-55805 Supplier Medchemexpress LLC Supplier No. HY14536A1G
Methylene blue is an inhibitor of guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). It functions as a vasopressor and is frequently employed as a dye in various medical procedures. This compound is a REDOX cycling compound capable of crossing the blood-brain barrier and has shown to reduce cerebral edema, attenuate microglial activation, and diminish neuroinflammation.
  • Inhibits guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS)
  • Functions as a vasopressor
  • Employed as a dye in medical procedures
  • Influences the nitric oxide synthase/guanylate cyclase signaling pathway
  • REDOX cycling compound
  • Capable of crossing the blood-brain barrier
  • Acts as a Tau aggregation inhibitor
  • Reduces cerebral edema
  • Attenuates microglial activation
  • Diminishes neuroinflammation