Learn More
Medchemexpress LLC Methylene blue | 61-73-4 | 95.0% | C16H18ClN3S | 10 G
SDP
Catalog No. 5000354400
Encompass_Preferred
Methylene blue is a guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS) inhibitor. It acts as a vasopressor and is frequently utilized as a dye in various medical procedures. This compound reduces prepulse inhibition through the nitric oxide synthase/guanylate cyclase signaling pathway.
  • REDOX cycling compound capable of crossing the blood-brain barrier.
  • Tau aggregation inhibitor.
  • Reduces cerebral edema.
  • Attenuates microglial activation.
  • Decreases neuroinflammation.
Catalog No. 50-003-54400 Supplier Medchemexpress LLC Supplier No. HY14536A10G
Add to Cart
Add to Cart
Methylene blue is a guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS) inhibitor. It acts as a vasopressor and is frequently utilized as a dye in various medical procedures. This compound reduces prepulse inhibition through the nitric oxide synthase/guanylate cyclase signaling pathway.
  • REDOX cycling compound capable of crossing the blood-brain barrier.
  • Tau aggregation inhibitor.
  • Reduces cerebral edema.
  • Attenuates microglial activation.
  • Decreases neuroinflammation.