Learn More
Medchemexpress LLC Methylene blue | 61-73-4 | 95.0% | C16H18ClN3S | 500 MG
SDP
Catalog No. 5000354911
Encompass_Preferred
Methylene blue is a chemical compound frequently used as a dye in medical procedures and functions as a vasopressor. It acts as an inhibitor for guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). Methylene blue influences the nitric oxide synthase/guanylate cyclase signaling pathway and is capable of crossing the blood-brain barrier. It also functions as a Tau aggregation inhibitor, demonstrating potential in reducing cerebral edema, attenuating microglial activation, and diminishing neuroinflammation.
  • Inhibits sGC, MAO-A, and NOS pathways
  • Used as a vasopressor and medical dye
  • Influences nitric oxide synthase/guanylate cyclase signaling
  • Crosses the blood-brain barrier
  • Functions as a Tau aggregation inhibitor
  • Reduces cerebral edema and neuroinflammation
Catalog No. 50-003-54911 Supplier Medchemexpress LLC Supplier No. HY14536A500MG
Methylene blue is a chemical compound frequently used as a dye in medical procedures and functions as a vasopressor. It acts as an inhibitor for guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). Methylene blue influences the nitric oxide synthase/guanylate cyclase signaling pathway and is capable of crossing the blood-brain barrier. It also functions as a Tau aggregation inhibitor, demonstrating potential in reducing cerebral edema, attenuating microglial activation, and diminishing neuroinflammation.
  • Inhibits sGC, MAO-A, and NOS pathways
  • Used as a vasopressor and medical dye
  • Influences nitric oxide synthase/guanylate cyclase signaling
  • Crosses the blood-brain barrier
  • Functions as a Tau aggregation inhibitor
  • Reduces cerebral edema and neuroinflammation