Learn More
Medchemexpress LLC Methylene blue | 61-73-4 | MFCD00012111 | 95.0% | C16H18ClN3S | 100 MG
SDP
Catalog No. 5000354399
Encompass_Preferred
Methylene blue is an inhibitor of guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). It acts as a vasopressor and is commonly used as a dye in various medical procedures. This REDOX cycling compound is capable of crossing the blood-brain barrier and is also a Tau aggregation inhibitor. It helps reduce cerebral edema, mitigate microglial activation, and decrease neuroinflammation.
  • Inhibits guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS)
  • Acts as a vasopressor
  • Commonly used as a dye in medical procedures
  • Impacts prepulse inhibition through the nitric oxide synthase/guanylate cyclase signaling pathway
  • Functions as a REDOX cycling compound
  • Capable of crossing the blood-brain barrier
  • Acts as a Tau aggregation inhibitor
  • Helps reduce cerebral edema
  • Mitigates microglial activation
  • Decreases neuroinflammation
Catalog No. 50-003-54399 Supplier Medchemexpress LLC Supplier No. HY14536A100MG
Methylene blue is an inhibitor of guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS). It acts as a vasopressor and is commonly used as a dye in various medical procedures. This REDOX cycling compound is capable of crossing the blood-brain barrier and is also a Tau aggregation inhibitor. It helps reduce cerebral edema, mitigate microglial activation, and decrease neuroinflammation.
  • Inhibits guanylyl cyclase (sGC), monoamine oxidase A (MAO-A), and NO synthase (NOS)
  • Acts as a vasopressor
  • Commonly used as a dye in medical procedures
  • Impacts prepulse inhibition through the nitric oxide synthase/guanylate cyclase signaling pathway
  • Functions as a REDOX cycling compound
  • Capable of crossing the blood-brain barrier
  • Acts as a Tau aggregation inhibitor
  • Helps reduce cerebral edema
  • Mitigates microglial activation
  • Decreases neuroinflammation