Learn More
Electron Microscopy Sciences Methylene Blue Azur B Solution 500 ML
SDP
Catalog No. 5031705
Encompass_Preferred
Used in Thomas Method for Malarial Parasites, Thomas (1953). References: Thomas, J.T. Stain Techn., 28:311-312, 1953 and Luna, L.G. (ed.) Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, New York, P.129, c.1968
Catalog No. 50-317-05 Supplier Electron Microscopy Sciences Supplier No. 2615502
Used in Thomas Method for Malarial Parasites, Thomas (1953). References: Thomas, J.T. Stain Techn., 28:311-312, 1953 and Luna, L.G. (ed.) Manual of Histologic Staining Methods of the AFIP, 3rd Ed. McGraw-Hill, New York, P.129, c.1968