Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Chloride Indicator Solution, Reagents

Catalog No. 01336763
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-763 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-763 Supplier Reagents Holdings Llc Supplier No. C6110400100C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylene Blue Chloride Indicator Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methylene Blue Chloride Indicator Solution
CAS 7732-18-5, 64-17-5, 67-56-1
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 0.9573
Molecular Formula C16H18ClN3S/H2O
Color Blue
Synonym 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015
Quantity 100 mL
Physical Form Liquid
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.