Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagents

Catalog No. 01336760
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-760 25 g
01-336-761 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-760 Supplier Reagents Holdings Llc Supplier No. C611050025C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methylene Blue Chloride Trihydrate (Basic Blue 9)
CAS 7220-79-3
Packaging Glass Bottle
Specific Gravity 1.0001
Molecular Formula C16H18ClN3S, 3H2O
Color Blue/Green
Synonym 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015
Quantity 25 g
Physical Form Solid
Molecular Weight (g/mol) 373.8981
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.