Learn More
Bioworld Loeffler's Alkaline Methylene Blue (Methylene Blue Counterstain), 100 mL
Catalog No. 50199226
Encompass_Preferred
Loeffler's Alkaline Methylene Blue (Methylene Blue Counterstain) can be used as a counter stain for Carbol-Fuchsin staining.The product is supplied as a two-component reagent in a ratio 1:3 (Dye:Base).Mix 1 part Solution A (MethyleneBlue) with 3 part Solution B (Dilute KOH).
Catalog No. 50-199-226 Supplier Bioworld Supplier No. 217700261
Loeffler's Alkaline Methylene Blue (Methylene Blue Counterstain) can be used as a counter stain for Carbol-Fuchsin staining.The product is supplied as a two-component reagent in a ratio 1:3 (Dye:Base).Mix 1 part Solution A (MethyleneBlue) with 3 part Solution B (Dilute KOH).