Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Methylene Blue (Loeffler's)
SCI_ED

Catalog No. S25432 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25432 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25432

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 131-69-1 , 122965-43-9 , 131-58-3 , 7732-18-5
Molecular Formula C16H14N2O6S
Molecular Weight (g/mol) 362.36
MDL Number MFCD00020275
InChI Key SNWQKAWITMVCQW-UHFFFAOYSA-N
PubChem CID 8574
IUPAC Name 2-{[4-(acetamidosulfonyl)phenyl]carbamoyl}benzoic acid
SMILES CC(=O)NS(=O)(=O)C1=CC=C(NC(=O)C2=CC=CC=C2C(O)=O)C=C1

Specifications

Assay Percent Range 30%v/v
Packaging Solution
Density 0.9g/mL
Quantity 500 mL
Chemical Name or Material Methylene Blue (Loeffler's)
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.