Learn More
Description
Certified for use in Mallory's phloxine-methylene blue stain; Levine and Black stain for bacteria in milk; Wright stain for blood smears.
Specifications
Specifications
| Chemical Name or Material | Methylene blue |
| CAS | 122965-43-9 |
| Color | Dark Green |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 319.85 g/mol |
| Percent Purity | ≥82% |
| Grade | Certified Biological Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.