Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Solution, Certified, ENG Scientific

Catalog No. ES6517
Change view
Click to view available options
Quantity:
2 x 250 mL
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6517 1 gal.
ES6515 2 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES6517 Supplier Eng Scientific Inc Supplier No. 6517
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ENG Scientific Certified Methylene Blue Solution is ideal for staining acid-fast bacteria.

  • Use with Acid-fast stain kit
  • Modification of the Kinyoun Method
  • Solution III - Loeffler's Methylene Blue

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Loeffler's Methylene Blue
PubChem CID 104827
IUPAC Name 3,7-bis(dimethylamino)-5λ4-phenothiazin-5-ylium trihydrate chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Packaging Bottle
Color Blue
Quantity 1 gal.
Chemical Name or Material Methylene Blue Solution

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.