Learn More
Description
Methylene blue is used to stain animal cells to make their nuclei more observable. It is a component of staining media such as Wright's stain and Jenner's stain. Methylene blue, also known as methylthioninium chloride, is a photoactive phenothiazine dye. It is a small molecular weight dye, which is effective against Gram positive and Gram negative bacteria.
Specifications
Specifications
| Chemical Name or Material | Methylene Blue |
| CAS | 61-73-4 |
| Color | Blue |
| Quantity | 250 mL |
| Molecular Weight (g/mol) | 319.85 g/mol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.