Learn More
Newcomer Supply Methylene Blue Stain 0.14%, Alcoholic, 250 ml
SDP
Catalog No. NC1751766
Encompass
Use: AFB Stock Counterstain for Kinyoun. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC1751766 Supplier Newcomer Supply Supplier No. 12401A
Use: AFB Stock Counterstain for Kinyoun. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.