Learn More
Medchemexpress LLC HY-B1359 100mg Medchemexpress, Methylene blue (trihydrate) CAS:7220-79-3 Purity:>98%
Supplier: Medchemexpress LLC HYB1359100MG
Medchemexpress, HY-B1359 100mg Methylene blue (trihydrate) CAS:7220-79-3 Methylene blue trihydrate (C.I. Basic Blue 9 trihydrate) is a guanylyl cyclase (sGC), monoamine oxidase A (MAO-A) and NO synthase (NOS) inhibitor. Methylene blue trihydrate is a vasopressor and is often used as a dye in several medical procedures. Methylene blue trihydrate has antinociception, antimalarial, antidepressant and anxiolytic activity effects. Methylene Blue trihydrate has the potential for methemoglobinemias, neurodegenerative disorders and ifosfamide-induced encephalopathytreatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.