Learn More
Alkali Scientific Methylene Blue Trihydrate, 500 G
SDP
Catalog No. 5000660519
Encompass_Preferred
Methylene Blue Trihydrate (500g) is an essential biological staining dye used for microscopic analysis in biological and microbiological research. This dye is particularly effective for staining living cells, bacteria, and tissue samples. Methylene Blue provides clear visibility of cellular structures and is an excellent stain for differentiating between viable and non-viable cells in vital staining procedures. The 500g size is perfect for high-throughput laboratories that require large quantities of this dye for routine or specialized staining protocols. It is used in clinical diagnostics, histology, and other research applications that require precise staining for cell and tissue analysis.
Catalog No. 50-006-60519 Supplier Alkali Scientific Supplier No. YLE003CP
Add to Cart
Add to Cart
Methylene Blue Trihydrate (500g) is an essential biological staining dye used for microscopic analysis in biological and microbiological research. This dye is particularly effective for staining living cells, bacteria, and tissue samples. Methylene Blue provides clear visibility of cellular structures and is an excellent stain for differentiating between viable and non-viable cells in vital staining procedures. The 500g size is perfect for high-throughput laboratories that require large quantities of this dye for routine or specialized staining protocols. It is used in clinical diagnostics, histology, and other research applications that require precise staining for cell and tissue analysis.