Learn More
Selleck Chemical LLC Methylene Blue trihydrate E4805-1G
SDP
Catalog No. 5000042501
Encompass_Preferred
Methylene blue trihydrate (C I Basic Blue 9 trihydrate) acts as an inhibitor of soluble guanylyl cyclase (sGC) monoamine oxidase A (MAO-A) and NO synthase (NOS) Methylene blue trihydrate exhibits antimalarial antidepressant and anxiolytic activity effects Methylene Blue trihydrate has potential applications in treating methemoglobinemia neurodegenerative disorders and ifosfamide-induced encephalopathy
Catalog No. 50-000-42501 Supplier Selleck Chemical LLC Supplier No. E48051G
Methylene blue trihydrate (C I Basic Blue 9 trihydrate) acts as an inhibitor of soluble guanylyl cyclase (sGC) monoamine oxidase A (MAO-A) and NO synthase (NOS) Methylene blue trihydrate exhibits antimalarial antidepressant and anxiolytic activity effects Methylene Blue trihydrate has potential applications in treating methemoglobinemia neurodegenerative disorders and ifosfamide-induced encephalopathy