Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene chloride, ≥99.8%, Ultra Resi-analyzed, J.T. Baker™

Catalog No. 02004651 Shop All Avantor Products
Click to view available options
Quantity:
4 L
Packaging:
Glass
This item is not returnable. View return policy

Catalog No. 02-004-651

Supplier: Avantor 926403

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -95°C
Color Colorless
Boiling Point 39.75°C
Quantity 4 L
Packaging Glass
Physical Form Liquid
Specific Gravity 1L = 1.32kg
Formula Weight 84.93
Percent Purity ≥99.8%
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Methylene chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.