Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene chloride, ≥99.8%, Ultra Resi-analyzed, J.T. Baker™

Catalog No. 02003395 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-395 1 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-003-395 Supplier Avantor Supplier No. 926402
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Organic Residue Analysis

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -95°C
Color Colorless
Quantity 1 L
Packaging Narrow Mouth Amber Glass Bottle
Physical Form Liquid
Specific Gravity 1L = 1.32kg
Boiling Point 39.75°C
Formula Weight 84.93
Percent Purity ≥99.8%
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Methylene chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.