Learn More
Cambridge Isotope Laboratories Methylene Chloride-d2 (d, 99.9%), 1665-00-5, MFCD00000882, 25 g
Catalog No. NC9773213
Encompass_Preferred
Chemical Formula: CD2Cl2, MW: 86.94, 99.8%, Synonyms: Dichloromethane; Methylene dichloride.
This item is not returnable. View return policy
Catalog No. NC9773213 Supplier Cambridge Isotope Laboratories Supplier No. DLM2325
Chemical Formula: CD2Cl2, MW: 86.94, 99.8%, Synonyms: Dichloromethane; Methylene dichloride.