Learn More
Methylene Chloride, Hydrocarbon Stabilized, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| CAS Max % | 100% |
| Boiling Point | 39.8°C/103.6°F |
| Packaging | Amber Glass Bottle |
| Quantity | 4 x 4 L |
| UN Number | UN1593 |
| Residue after Evaporation | 4 ppm |
| Percent Purity | 99.9% |
| Grade | HPLC |
| Physical Form | Liquid |
| Chemical Name or Material | Methylene Chloride, Hydrocarbon Stabilized |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.