Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, NF, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604514 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-514 500 mL Amber Glass Bottle
18-604-513 100 mL Amber Glass Bottle
18-604-516 1 L Amber Glass Bottle
18-604-515 6 x 500 mL Amber Glass Bottle
18-604-517 4 L Amber Glass Bottle
18-604-518 6 x 1 L Amber Glass Bottle
18-604-519 4 x 4 L Amber Glass Bottle
18-604-520 20 L Steel Can
18-604-521 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-604-514 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1266500MLGL
Only null left
Add to Cart
Add to Cart

M1266, 75-09-2, CH2Cl2

Spectrum™ Chemical Methylene Chloride, NF is used as a carrier for pharmaceutical tablet coatings. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
UN Number UN1593
Boiling Point 39.8°C
Percent Purity 99%
Grade NF
Chemical Name or Material Methylene Chloride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.