Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, NF, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604521 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-521 200 L Drum
18-604-513 100 mL Amber Glass Bottle
18-604-514 500 mL Amber Glass Bottle
18-604-516 1 L Amber Glass Bottle
18-604-515 6 x 500 mL Amber Glass Bottle
18-604-517 4 L Amber Glass Bottle
18-604-518 6 x 1 L Amber Glass Bottle
18-604-519 4 x 4 L Amber Glass Bottle
18-604-520 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 18-604-521 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1266200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

M1266, 75-09-2, CH2Cl2

Spectrum™ Chemical Methylene Chloride, NF is used as a carrier for pharmaceutical tablet coatings. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N

Specifications

Quantity 200 L
Packaging Drum
UN Number UN1593
Boiling Point 39.8°C
Grade NF
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.