Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical PROMO

Catalog No. D1514FB25 Shop All Fisherbrand Products
Change view
Click to view available options
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
D1514FB25 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1514FB25

missing translation for 'validMainframeMsg'

1 L Dichloromethane, for residue analysis. Distol™ stabilized with amylene.

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Impurity profile is also reported using gas chromatography and three different detectors (ECD, FID, and MS) to demonstrate suitable for different analyses
  • Contains approximately 15 to 200 ppm amylene as preservative
  • Also meets ACS specifications
  • Intended for research use only
  • CAS: 75-09-2, CH2Cl2, 84.93 g/mol
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Quantity 4 L
Identification Pass Test
Packaging Amber Soda Lime Glass
Physical Form Liquid
Assay Percent Range 99.9% min by GC
Boiling Point 39°C
Formula Weight 84.93 amu
Grade High Purity Mobile Phase for HPLC and/or Extraction Solvent for GC Applications
Chemical Name or Material Methylene Chloride
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.