Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017852 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7852 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-7852 Supplier MilliporeSigma Supelco Supplier No. PHR15573X1.2ML
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Ignition Point 1223°F
CAS 75-09-2
Vapor Pressure 24.45 psi (55°C)
Vapor Density 2.9 (vs. air)
Density 1.325 g/mL (at 25°C)
Linear Formula CH2 Cl2
MDL Number MFCD00000881
Melting Point -97°C
Boiling Point 39.8°C to 40°C
Synonym Dichloromethane; Methylene chloride
Type Certified Reference Material
Quantity 3 x 1.2 mL
Formula Weight 84.93
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.