Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride, PHOTREX™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

75-09-2
CH2Cl2
84.927
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
CHEBI:15767

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Spectrophotometry
CAS 75-09-2
Molecular Formula CH2Cl2
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES C(Cl)Cl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.927
PubChem CID 6344
ChEBI CHEBI:15767
Formula Weight 84.93
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.32kg
Quantity 500mL
Chemical Name or Material Methylene Chloride
Grade PHOTREX™ Reagent
Show More Show Less