Learn More
Electron Microscopy Sciences Methylene Chloride (Plastisolve) 1 QT
SDP
Catalog No. 5027888
Encompass_Preferred
(Dichloromethane; Plastisolve)
CAS #75-09-2 CH₂Cl₂
Good for removing cured or uncured epoxy resins.
Catalog No. 50-278-88 Supplier Electron Microscopy Sciences Supplier No. 19600
(Dichloromethane; Plastisolve)
CAS #75-09-2 CH₂Cl₂
Good for removing cured or uncured epoxy resins.