Learn More
Electron Microscopy Sciences Methylene Chloride (Plastisolve) 1 QT
SDP
Supplier:  Electron Microscopy Sciences 19610
Encompass_Preferred
(Dichloromethane; Plastisolve)
CAS #75-09-2 CH₂Cl₂
Good for removing cured or uncured epoxy resins.
Catalog No. 50-278-87