Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene diurethane, 98+%, Thermo Scientific™

Catalog No. p-7043342
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0031814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0031814 Supplier Thermo Scientific Chemicals Supplier No. L0031814

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3693-53-6
Molecular Formula C7H14N2O4
Molecular Weight (g/mol) 190.20
MDL Number MFCD00015147
InChI Key QSPOJWMZYOMAEI-UHFFFAOYSA-N
Synonym methylene diurethane, methylenediurethane, methylene diurethan, diethyl methylenedicarbamate, methylene bisurethane, diethyl methylenebiscarbamate, bis n-ethoxycarbonylamino methane, carbamic acid, methylenedi-, diethyl ester, carbamic acid, methylenebis-, diethyl ester, n,n-methylene-bis ethyl carbamate
PubChem CID 19407
IUPAC Name ethyl N-[(ethoxycarbonylamino)methyl]carbamate
SMILES CCOC(=O)NCNC(=O)OCC

Specifications

Melting Point 128°C to 132°C
Beilstein 1812852
Quantity 25 g
Formula Weight 190.2
Percent Purity ≥98%
Chemical Name or Material Methylene diurethane
EINECSNumber 223-011-4
RTECSNumber FC2200000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.