Learn More
Medchemexpress LLC Methylene violet 3RAX | 4569-86-2 | MFCD00013433 | 95.5% | 378.9 g/mol | C22H23ClN4 | 5 G
SDP
Catalog No. 5000282978
Encompass_Preferred
Methylene Violet 3RAX is a phenazine-class dye used to stain mitochondria in cells and to highlight cellular structures in histological and microbiological preparations. It interacts with nucleic acids and is used in research staining protocols where mitochondrial visualization and cellular contrast are required. The reagent is supplied as a solid with reported high purity for laboratory use.
  • Phenazine-class dye for mitochondrial staining.
  • Interacts with DNA to enhance cellular contrast in assays.
  • High purity suitable for research applications.
  • Available as a solid reagent for easy handling and storage.
  • Soluble in water and common laboratory solvents for versatile protocols.
  • Applicable to histology, microbiology, and cell imaging workflows.
Catalog No. 50-002-82978 Supplier Medchemexpress LLC Supplier No. HYW1302365G
Methylene Violet 3RAX is a phenazine-class dye used to stain mitochondria in cells and to highlight cellular structures in histological and microbiological preparations. It interacts with nucleic acids and is used in research staining protocols where mitochondrial visualization and cellular contrast are required. The reagent is supplied as a solid with reported high purity for laboratory use.
  • Phenazine-class dye for mitochondrial staining.
  • Interacts with DNA to enhance cellular contrast in assays.
  • High purity suitable for research applications.
  • Available as a solid reagent for easy handling and storage.
  • Soluble in water and common laboratory solvents for versatile protocols.
  • Applicable to histology, microbiology, and cell imaging workflows.