Learn More
Medchemexpress LLC Methylene violet 3RAX | 4569-86-2 | MFCD00013433 | 95.5% | 378.90 g/mol | C22H23ClN4 | 10 G
SDP
Catalog No. 5000283678
Encompass_Preferred
Methylene violet 3RAX is a phenazine-class dye (CAS 4569-86-2) used as a mitochondrial stain and as a photosensitizer in research applications such as photodynamic therapy. It has reported effects on DNA structure and shows inhibitory activity against acetylcholinesterase and butyrylcholinesterase. The material is supplied as a solid with a typical purity of 95.5%.
  • Phenazine-class mitochondrial staining dye.
  • Useful as a photosensitizer for mitochondrial targeting studies.
  • Reported to affect DNA structure, enabling mechanistic studies.
  • Exhibits inhibitory activity against AChE and BChE for biochemical assays.
  • Supplied as a solid with typical purity of 95.5% and multiple package sizes, including 10 G.
Catalog No. 50-002-83678 Supplier Medchemexpress LLC Supplier No. HYW13023610G
Methylene violet 3RAX is a phenazine-class dye (CAS 4569-86-2) used as a mitochondrial stain and as a photosensitizer in research applications such as photodynamic therapy. It has reported effects on DNA structure and shows inhibitory activity against acetylcholinesterase and butyrylcholinesterase. The material is supplied as a solid with a typical purity of 95.5%.
  • Phenazine-class mitochondrial staining dye.
  • Useful as a photosensitizer for mitochondrial targeting studies.
  • Reported to affect DNA structure, enabling mechanistic studies.
  • Exhibits inhibitory activity against AChE and BChE for biochemical assays.
  • Supplied as a solid with typical purity of 95.5% and multiple package sizes, including 10 G.