Learn More
Medchemexpress LLC Methylene violet 3RAX | 4569-86-2 | MFCD00013433 | 95.5% | 378.90 g/mol | C22H23ClN4 | 500 MG
SDP
Catalog No. 5000283558
Encompass_Preferred
Methylene Violet 3RAX is a phenazine-class dye provided as a green to dark green powder for research staining applications. It is primarily used for mitochondrial and histological staining and is supplied with a reported purity suitable for laboratory use. Store protected from light and follow recommended solvent storage conditions for extended stability.
  • Phenazine-class dye suitable for mitochondrial and histological staining
  • Green to dark green powder form, convenient for laboratory preparation
  • Supplied with reported purity of 95.5% for research use
  • Stable when stored protected from light at 4°C; follow solvent storage recommendations for long-term storage
  • Compatible with common staining protocols in cell biology and microbiology
Catalog No. 50-002-83558 Supplier Medchemexpress LLC Supplier No. HYW130236500MG
Methylene Violet 3RAX is a phenazine-class dye provided as a green to dark green powder for research staining applications. It is primarily used for mitochondrial and histological staining and is supplied with a reported purity suitable for laboratory use. Store protected from light and follow recommended solvent storage conditions for extended stability.
  • Phenazine-class dye suitable for mitochondrial and histological staining
  • Green to dark green powder form, convenient for laboratory preparation
  • Supplied with reported purity of 95.5% for research use
  • Stable when stored protected from light at 4°C; follow solvent storage recommendations for long-term storage
  • Compatible with common staining protocols in cell biology and microbiology