Learn More
Medchemexpress LLC Methylene violet 3RAX | 4569-86-2 | MFCD00013433 | 95.5% | 378.91 g/mol | C22H23N4Cl | 25 G
SDP
Catalog No. 5000283278
Encompass_Preferred
Methylene Violet 3RAX is a phenazine-class dye used for mitochondrial staining and photodynamic therapy research. It interacts with DNA, produces reactive singlet oxygen, and exhibits inhibitory activity against acetylcholinesterase and butyrylcholinesterase, making it useful for mitochondrial targeting studies and biochemical assays.
  • Phenazine dye for mitochondrial staining.
  • Generates reactive singlet oxygen for photodynamic studies.
  • Interacts with DNA and can alter nucleic acid structure.
  • Shows AChE and BChE inhibitory activity for biochemical assays.
  • Available as a solid powder with high reported purity.
Catalog No. 50-002-83278 Supplier Medchemexpress LLC Supplier No. HYW13023625G
Methylene Violet 3RAX is a phenazine-class dye used for mitochondrial staining and photodynamic therapy research. It interacts with DNA, produces reactive singlet oxygen, and exhibits inhibitory activity against acetylcholinesterase and butyrylcholinesterase, making it useful for mitochondrial targeting studies and biochemical assays.
  • Phenazine dye for mitochondrial staining.
  • Generates reactive singlet oxygen for photodynamic studies.
  • Interacts with DNA and can alter nucleic acid structure.
  • Shows AChE and BChE inhibitory activity for biochemical assays.
  • Available as a solid powder with high reported purity.