Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methylenediphosphonic acid

Catalog No. M95085G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M95085G 5 g
M950810G 10 g
M95081G 1 g
M9508 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M95085G Supplier Sigma Aldrich Supplier No. M95085G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methylenediphosphonic acid

Specifications

CAS 1984-15-2
Melting Point 197°C to 199°C (lit.)
Molecular Formula CH6O6P2
Linear Formula CH2[P(O)(OH)2]2
MDL Number MFCD00002139
Quantity 5 g
Synonym MDP; Medronic acid
Molecular Weight (g/mol) 176
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.