Learn More
Medchemexpress LLC Methylenephosphonic acid CD73 inhibitor | 2216764-29-1 | 99.2% | 463.81 g·mol⁻¹ | C16H23ClN5O7P | 5 MG
SDP
Catalog No. 5000262482
Encompass_Preferred
CD73-IN-4 is a potent, selective methylenephosphonic acid inhibitor of ecto-5'-nucleotidase (CD73) with low-nanomolar activity against human CD73. It is supplied as a research reagent for biochemical and cell-based studies, with reported high purity and multiple pack size options. Molecular formula C16H23ClN5O7P; molecular weight 463.81 g·mol⁻¹.
Catalog No. 50-002-62482 Supplier Medchemexpress LLC Supplier No. HY1319675MG
CD73-IN-4 is a potent, selective methylenephosphonic acid inhibitor of ecto-5'-nucleotidase (CD73) with low-nanomolar activity against human CD73. It is supplied as a research reagent for biochemical and cell-based studies, with reported high purity and multiple pack size options. Molecular formula C16H23ClN5O7P; molecular weight 463.81 g·mol⁻¹.