Learn More
Apexbio Technology LLC Methylergometrine maleate 57432-61-8 250mg
SDP
Catalog No. 5000088987
Encompass_Preferred
Methylergometrine maleate (CAS 57432-61-8) is a semi-synthetic ergot alkaloid and the active metabolite of methylergometrine It exhibits potent uterotonic and vasoconstrictive effects through partial agonism and antagonism at serotonergic and dopaminergic receptors In laboratory studies methylergometrine maleate has demonstrated receptor antagonistic activity with an antagonistic value of 9 6 Its pharmacological properties support its use in the prevention and management of postpartum hemorrhage as well as its investigation in migraine research due to its dopaminergic modulation and serotonin receptor interactions
Catalog No. 50-000-88987 Supplier Apexbio Technology LLC Supplier No. B6347250
Methylergometrine maleate (CAS 57432-61-8) is a semi-synthetic ergot alkaloid and the active metabolite of methylergometrine It exhibits potent uterotonic and vasoconstrictive effects through partial agonism and antagonism at serotonergic and dopaminergic receptors In laboratory studies methylergometrine maleate has demonstrated receptor antagonistic activity with an antagonistic value of 9 6 Its pharmacological properties support its use in the prevention and management of postpartum hemorrhage as well as its investigation in migraine research due to its dopaminergic modulation and serotonin receptor interactions