Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylfluorone Hemisulfate [for Detection of Sb,Ge,Mo,Sn], TCI America™
SDP

Catalog No. M06781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M06781G Supplier TCI America Supplier No. M06781G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Formula C14H10O5
MDL Number MFCD00022274
Synonym 9-Methyl-2,3,7-trihydroxy-6-fluorone Hemisulfate

Specifications

Quantity 1 g
Formula Weight 307.27
Physical Form Crystalline Powder
Chemical Name or Material Methylfluorone Hemisulfate [for Detection of Sb,Ge,Mo,Sn]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.