Learn More
Medchemexpress LLC Methylglyoxal-bis(guanylhydrazone) | 459-86-9 | MFCD00067519 | ≥98.0% | 184.20 g·mol⁻¹ | C5H12N8 | 50 MG
SDP
Catalog No. 5000240386
Encompass_Preferred
Mitoguazone (methylglyoxal-bis(guanylhydrazone)) is a small-molecule research reagent that competitively inhibits S-adenosyl-methionine decarboxylase (SAMDC) and disrupts polyamine biosynthesis. It is used in biochemical and cellular studies to probe polyamine-dependent processes, mitochondrial effects, and apoptosis. Chemical identifiers include CAS 459-86-9, formula C5H12N8, and molecular weight 184.20 g·mol⁻¹.
  • Competitive inhibitor of S-adenosyl-methionine decarboxylase (SAMDC).
  • Disrupts polyamine biosynthesis for pathway and apoptosis research.
  • Reported to be brain-penetrant and useful in cellular assays.
  • Characterized chemical identifiers: CAS 459-86-9, formula C5H12N8, MW 184.20 g·mol⁻¹.
  • Recommended storage sealed and away from moisture; in solvent: -80°C long-term, -20°C short-term.
  • Reported supplier purity commonly ≥98% for research-grade material.
Catalog No. 50-002-40386 Supplier Medchemexpress LLC Supplier No. HY10663450MG
Mitoguazone (methylglyoxal-bis(guanylhydrazone)) is a small-molecule research reagent that competitively inhibits S-adenosyl-methionine decarboxylase (SAMDC) and disrupts polyamine biosynthesis. It is used in biochemical and cellular studies to probe polyamine-dependent processes, mitochondrial effects, and apoptosis. Chemical identifiers include CAS 459-86-9, formula C5H12N8, and molecular weight 184.20 g·mol⁻¹.
  • Competitive inhibitor of S-adenosyl-methionine decarboxylase (SAMDC).
  • Disrupts polyamine biosynthesis for pathway and apoptosis research.
  • Reported to be brain-penetrant and useful in cellular assays.
  • Characterized chemical identifiers: CAS 459-86-9, formula C5H12N8, MW 184.20 g·mol⁻¹.
  • Recommended storage sealed and away from moisture; in solvent: -80°C long-term, -20°C short-term.
  • Reported supplier purity commonly ≥98% for research-grade material.