Learn More
Medchemexpress LLC Methylhexanamine hydrochloride | 13803-74-2 | MFCD09038354 | 151.68 g/mol | C7H18ClN | 5 MG
SDP
Catalog No. 5000300666
Encompass_Preferred
Methylhexanamine hydrochloride is an aliphatic amine and adrenergic receptor agonist (CAS 13803-74-2) used in research as a vasoconstrictor; it was historically marketed as a nasal decongestant. This reagent is supplied for laboratory research use and is not intended for human therapeutic use.
  • Aliphatic amine and adrenergic receptor agonist
  • Acts as a vasoconstrictor and was used as a nasal decongestant
  • Provided for research use only; not for human administration
  • Molecular formula C7H18ClN; molecular weight 151.68 g/mol
  • Available in small milligram pack sizes suitable for assay development
  • Data sheet and handling instructions available for laboratory safety
Catalog No. 50-003-00666 Supplier Medchemexpress LLC Supplier No. HYW0155935MG
Methylhexanamine hydrochloride is an aliphatic amine and adrenergic receptor agonist (CAS 13803-74-2) used in research as a vasoconstrictor; it was historically marketed as a nasal decongestant. This reagent is supplied for laboratory research use and is not intended for human therapeutic use.
  • Aliphatic amine and adrenergic receptor agonist
  • Acts as a vasoconstrictor and was used as a nasal decongestant
  • Provided for research use only; not for human administration
  • Molecular formula C7H18ClN; molecular weight 151.68 g/mol
  • Available in small milligram pack sizes suitable for assay development
  • Data sheet and handling instructions available for laboratory safety