Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyliminodiacetic Acid, MP Biomedicals

Catalog No. ICN202482 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202482 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202482 Supplier MP Biomedicals, Inc Supplier No. 0520248205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4408-64-4
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
InChI Key XWSGEVNYFYKXCP-UHFFFAOYSA-N
Synonym n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
PubChem CID 20441
IUPAC Name 2-[carboxymethyl(methyl)amino]acetic acid
SMILES CN(CC(=O)O)CC(=O)O

Specifications

Quantity 10 g
Formula Weight 147.13
Chemical Name or Material Methyliminodiacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.