Learn More
Medchemexpress LLC Methylisothiazolinone hydrochloride | 26172-54-3 | 99.4% | 151.61 | 25 G
SDP
Catalog No. 5000451787
Encompass_Preferred
Methylisothiazolinone hydrochloride is a bacterial and fungal inhibitor and preservative, also acting as a sensitizer. It activates matrix metalloproteinases (MMPs) in human bronchial epithelial cells, leading to apoptosis and inflammatory responses. This compound can also promote the development of atopic dermatitis in mice by disrupting Th2/Th17 related immune responses and cause mitochondrial damage in the endothelium of rat cerebral blood vessels.
  • Acts as a bacterial and fungal inhibitor and preservative
  • Induces cell apoptosis and inflammatory response
  • Activates matrix metalloproteinases (MMPs)
  • Promotes atopic dermatitis development in mice
  • Causes mitochondrial damage in endothelium of rat cerebral blood vessels
  • Decreases cell viability in bronchial epithelial cells
  • Increases susceptibility of rat cerebral vascular endothelium to pathological stimuli
Catalog No. 50-004-51787 Supplier Medchemexpress LLC Supplier No. 25GHYW010243
Methylisothiazolinone hydrochloride is a bacterial and fungal inhibitor and preservative, also acting as a sensitizer. It activates matrix metalloproteinases (MMPs) in human bronchial epithelial cells, leading to apoptosis and inflammatory responses. This compound can also promote the development of atopic dermatitis in mice by disrupting Th2/Th17 related immune responses and cause mitochondrial damage in the endothelium of rat cerebral blood vessels.
  • Acts as a bacterial and fungal inhibitor and preservative
  • Induces cell apoptosis and inflammatory response
  • Activates matrix metalloproteinases (MMPs)
  • Promotes atopic dermatitis development in mice
  • Causes mitochondrial damage in endothelium of rat cerebral blood vessels
  • Decreases cell viability in bronchial epithelial cells
  • Increases susceptibility of rat cerebral vascular endothelium to pathological stimuli