Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyllithium lithium bromide complex, 2.2M sol. in diethylether, AcroSeal™, Thermo Scientific™

Catalog No. AC181290700
Change view
Click to view available options
Quantity:
70 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC181290700 70 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC181290700 Supplier Thermo Scientific Chemicals Supplier No. COMPLEX
Request Bulk or Custom Formats

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH3Li·LiBr
MDL Number MFCD00044645
Synonym dilithium carbanide bromide, methyllithium lithium bromide complex solution, dilithium 1+ ion bromide methanide

Specifications

Color Colorless to Yellow
CAS 60-29-7
Density 0.81g/mL
Quantity 70 mL
Physical Form Solution
Linear Formula CH3Li·LiBr
Formula Weight 108.82
Percent Purity 9.0 to 11.0% (LiBr), 6.0 to 7.0% (LiCH3)
Chemical Name or Material Methyllithium lithium bromide complex
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Extremely flammable liquid and v
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas. Protect from moisture.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Product may develop some turbidity or precipitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.