Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyllithium Solution

Catalog No. 503081455 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
4 x 25 mL
800 mL
1 L
4 x 100 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1455 4 x 25 mL
19-734-31L 1 L
19-734-3800ML 800 mL
50-308-1453 100 mL
19-734-3100ML 4 x 100 mL
19-734-350ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Danger
Catalog No. 50-308-1455 Supplier Sigma Aldrich Supplier No. 1973434X25ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyllithium is an organolithium reagent mainly used for deprotonation and as a source of methyl anion. It is a strong base and a strong nucleophile.Methyllithium solution (1.6M in diethyl ether) has been used for the asymmetric allylic alkylation (AAA) of allylic electrophiles in the presence of a chiral copper catalyst. This protocol leads to the C-C bond formation of tertiary and quaternary stereogenic centers with high enantioselectivity. It can also be used for the synthesis of (-)-salsolidine by reacting with 6,7-dimethoxy-3,4-dihydroisoquinoline in the presence of (-)-sparteine as a chiral ligand.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 917-54-4
Density 0.732 g/mL at 25°C
Molecular Formula CH3Li
Linear Formula CH3Li
MDL Number MFCD00008253
Quantity 4 x 25 mL
Synonym Lithium methanide; MeLi
Molecular Weight (g/mol) 21.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.