Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyllithium Solution

Catalog No. 51433025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-433-025ML 4 x 25 mL
51-433-01L 1 L
51-433-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 51-433-025ML Supplier Sigma Aldrich Supplier No. 5143304X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyllithium solution ( 3.1 M in diethoxymethane) can undergo 1,2-addition to benzaldehyde catalyzed by a 2-(2-hydroxyaryl)alcohol (HAROL) ligand in the presence of titanium isopropoxide to form (S)-1-phenylethan-1-ol.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 917-54-4
Density 0.846 g/mL at 25°C
Molecular Formula CH3Li
Linear Formula CH3Li
MDL Number MFCD00008253
Quantity 4 x 25 mL
Synonym Lithium methanide; MeLi
Molecular Weight (g/mol) 21.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.