Learn More
Cayman Chemical Methyl-L-NIO (hydrochloride), Purity: ≥98%, CAS: 150403-96-6, 5 mg
SDP
Catalog No. 502888235
Encompass_Preferred
A competitive NOS inhibitor; more potent inhibitor of nNOS (Ki = 3.0 µM) than eNOS (Ki = 10.0 µM) or iNOS (Ki = 9.5 µM)
Catalog No. 50-288-8235 Supplier Cayman Chemical Supplier No. 100102525 MG
A competitive NOS inhibitor; more potent inhibitor of nNOS (Ki = 3.0 µM) than eNOS (Ki = 10.0 µM) or iNOS (Ki = 9.5 µM)