Learn More
Medchemexpress LLC HY-N2332A 50mg , Methyllycaconitine citrate CAS:351344-10-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2332A50MG
Encompass_Preferred
HY-N2332A 50mg Methyllycaconitine citrate CAS: 351344-10-0 Methyllycaconitine citrate is a specific antagonist of α7 neuronal nicotinic acetylcholine receptor (α7nAChR). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1191